LIF517189586 Life Chemicals Inc.

3-methyl-3-phenylbutanoic acid

  • MDL number MFCD00020492
  • Cas Number : 1010-48-6
  • Molecular Weight : 178.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart