LIF517189587 Life Chemicals Inc.

2-(chloromethyl)-5-(oxan-4-yl)-1,3,4-oxadiazole

  • MDL number MFCD17242174
  • Cas Number : 1247061-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart