LIF517189591 Life Chemicals Inc.

[4-(4-methylphenoxy)phenyl]methanamine hydrochloride

  • MDL number MFCD02089411
  • Cas Number : 262862-66-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart