LIF517189596 Life Chemicals Inc.

ethyl (2E)-4-aminobut-2-enoate; trifluoroacetic acid

  • MDL number MFCD27922553
  • Cas Number : 1218810-53-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart