LIF517189599 Life Chemicals Inc.

3-{[1,1'-biphenyl]-3-yl}propanoic acid

  • MDL number MFCD19319922
  • Cas Number : 404362-38-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart