LIF945291005 Life Chemicals Inc.

5-cyclopropyl-1-propyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD14609323
  • Cas Number : 1248463-84-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart