LIF945291008 Life Chemicals Inc.

cyclopropylurea

  • MDL number MFCD01569844
  • Cas Number : 19757-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart