LIF945291011 Life Chemicals Inc.

2,3-dichloro-5,6-diethylpyrazine

  • MDL number MFCD26407869
  • Cas Number : 1461705-47-4
  • Molecular Weight : 205.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart