LIF945291017 Life Chemicals Inc.

2,3-dihydro-1-benzofuran-3-carboxylic acid

  • MDL number MFCD08669486
  • Cas Number : 39891-55-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart