LIF945291030 Life Chemicals Inc.

3-chloro-5,6,7,8-tetrahydrocinnoline

  • MDL number MFCD16547518
  • Cas Number : 32078-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart