LIF945291031 Life Chemicals Inc.

2,2,6,6-tetrafluoromorpholine

  • MDL number MFCD09971734
  • Cas Number : 65502-95-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart