LIF945291044 Life Chemicals Inc.

5-(propan-2-yloxy)pyridine-3-carboxylic acid

  • MDL number MFCD14282014
  • Cas Number : 863507-81-7
  • Molecular Weight : 181.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart