LIF945291051 Life Chemicals Inc.

4-{[(tert-butoxy)carbonyl](methyl)amino}butanoic acid

  • MDL number MFCD09831984
  • Cas Number : 94994-39-5
  • Molecular Weight : 217.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart