LIF945291084 Life Chemicals Inc.

5-fluoro-2-(morpholin-4-yl)benzaldehyde

  • MDL number MFCD06656555
  • Cas Number : 883535-68-0
  • Molecular Weight : 209.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart