LIF945291090 Life Chemicals Inc.

(1-methylcyclobutyl)methanol

  • MDL number MFCD18072623
  • Cas Number : 38401-41-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart