LIF945291091 Life Chemicals Inc.

8-methoxy-3,4-dihydro-2H-1-benzopyran-3-carboxylic acid

  • MDL number MFCD04114622
  • Cas Number : 108088-19-3
  • Molecular Weight : 208.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart