LIF945291119 Life Chemicals Inc.

5-(chloromethyl)-3-cyclopropyl-1,2-oxazole

  • MDL number MFCD19234016
  • Cas Number : 127450-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart