LIF945291120 Life Chemicals Inc.

methyl 2-(3-amino-1H-pyrazol-1-yl)acetate

  • MDL number MFCD12149369
  • Cas Number : 1093967-39-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart