LIF945291121 Life Chemicals Inc.

8-fluoro-3-iodo-6-methylquinoline

  • MDL number MFCD27500866
  • Cas Number : 1592298-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart