LIF945291132 Life Chemicals Inc.

[3-(cyclopentyloxy)phenyl]methanamine

  • MDL number MFCD09729663
  • Cas Number : 953717-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart