LIF945291133 Life Chemicals Inc.

4-(chloromethyl)-5-methyl-1,3-oxazole

  • MDL number MFCD16989275
  • Cas Number : 1240612-11-6
  • Molecular Weight : 131.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart