LIF945291138 Life Chemicals Inc.

tert-butyl piperidine-3-carboxylate

  • MDL number MFCD11041017
  • Cas Number : 301180-05-2
  • Molecular Weight : 185.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart