LIF945291142 Life Chemicals Inc.

1-ethylcyclobutane-1-carboxylic acid

  • MDL number MFCD18253511
  • Cas Number : 150864-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart