LIF945291166 Life Chemicals Inc.

8-fluoro-3-iodo-5-nitroquinoline

  • MDL number MFCD19982608
  • Cas Number : 1334148-68-3
  • Molecular Weight : 318.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart