LIF945291175 Life Chemicals Inc.

tert-butyl 5-(hydroxymethyl)-2-azabicyclo[3.1.0]hexane-2-carboxylate

  • MDL number MFCD28133352
  • Cas Number : 1630855-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form oil
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart