LIF945291176 Life Chemicals Inc.

(5-methyl-1H-pyrazol-3-yl)methanol

  • MDL number MFCD00159583
  • Cas Number : 29004-73-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart