LIF945291188 Life Chemicals Inc.

3-(pyrrolidin-1-yl)propanenitrile

  • MDL number MFCD00014099
  • Cas Number : 26165-45-7
  • Molecular Weight : 124.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin UA
Availability details for
Quantity
  • Availability
  • Add to Cart