LIF945291190 Life Chemicals Inc.

3-[(3-bromophenyl)amino]propanenitrile

  • MDL number MFCD11164272
  • Cas Number : 1049118-66-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart