LIF945291193 Life Chemicals Inc.

2-(5-bromothiophen-2-yl)morpholine

  • MDL number MFCD12819882
  • Cas Number : 1178409-69-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart