LIF945291194 Life Chemicals Inc.

(2-ethoxypyridin-4-yl)methanol

  • MDL number MFCD14606409
  • Cas Number : 153928-58-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart