LIF945291198 Life Chemicals Inc.

2-(1H-pyrazol-1-yl)cyclohexan-1-ol

  • MDL number MFCD12765021

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart