LIF945291205 Life Chemicals Inc.

2-phenyl-1,3-thiazolidine

  • MDL number MFCD00128238
  • Cas Number : 4569-82-8
  • Molecular Weight : 165.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart