LIF945291206 Life Chemicals Inc.

2-(2-chlorophenyl)-1,3-thiazolidine

  • MDL number MFCD00222815
  • Cas Number : 67189-26-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart