LIF945291208 Life Chemicals Inc.

2-chloro-4-(ethylsulfanyl)pyrimidine

  • MDL number MFCD16712642
  • Cas Number : 1250161-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart