LIF945291213 Life Chemicals Inc.

1-(thiophen-3-yl)cyclopropan-1-ol

  • MDL number MFCD16840911
  • Cas Number : 888505-22-4
  • Molecular Weight : 140.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart