LIF945291222 Life Chemicals Inc.

7-chloro-2,5-dimethylpyrazolo[1,5-a]pyrimidine

  • MDL number MFCD11043774
  • Cas Number : 136549-13-8
  • Molecular Weight : 181.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart