LIF945291225 Life Chemicals Inc.

[(1S)-1-phenylethyl](propan-2-yl)amine

  • MDL number MFCD01862157
  • Cas Number : 19302-32-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart