LIF945291226 Life Chemicals Inc.

1,1,1-trifluoro-3-(1H-imidazol-1-yl)propan-2-ol

  • MDL number MFCD19602657
  • Cas Number : 1343763-62-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart