LIF945291235 Life Chemicals Inc.

2-chloro-4,5,6,7-tetrahydro-1,3-benzothiazole

  • MDL number MFCD08689787
  • Cas Number : 26846-98-0
  • Molecular Weight : 173.67

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart