LIF945291238 Life Chemicals Inc.

4-(pyridin-4-yl)pyrrolidin-2-one

  • MDL number MFCD19347216
  • Cas Number : 1268132-89-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart