LIF945291240 Life Chemicals Inc.

1-(bromomethyl)-1-methylcyclobutane

  • MDL number MFCD20627042
  • Cas Number : 98775-14-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart