LIF945291248 Life Chemicals Inc.

1-(pyridin-2-yl)but-3-en-1-ol

  • MDL number MFCD14653141
  • Cas Number : 17285-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart