LIF945291252 Life Chemicals Inc.

4-(1,3-thiazol-2-yl)butan-2-one

  • MDL number MFCD18813405
  • Cas Number : 933740-86-4
  • Molecular Weight : 155.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart