LIF945291253 Life Chemicals Inc.

1H-1,2,3-triazole-4-carbonitrile

  • MDL number MFCD00225621
  • Cas Number : 18755-49-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart