LIF945291261 Life Chemicals Inc.

(1-ethylcyclobutyl)methanol

  • MDL number MFCD12024521
  • Cas Number : 255721-49-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart