LIF945291264 Life Chemicals Inc.

2-amino-4-fluoro-5-methylbenzene-1-thiol

  • MDL number MFCD22376104
  • Cas Number : 1565977-92-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart