LIF945291265 Life Chemicals Inc.

ethyl 5-sulfanylthiophene-3-carboxylate

  • MDL number MFCD26842024
  • Cas Number : 1566962-85-3
  • Molecular Weight : 188.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form liquid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart