LIF945291270 Life Chemicals Inc.

6-chloro-3-cyclopropyl-1,2,3,4-tetrahydropyrimidine-2,4-dione

  • MDL number MFCD26867759
  • Cas Number : 1565120-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart