LIF945291272 Life Chemicals Inc.

N-propylthiomorpholine-4-carboxamide

  • MDL number MFCD28148567
  • Cas Number : 1600129-98-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart