LIF945291274 Life Chemicals Inc.

1-methoxycycloheptane-1-carboxylic acid

  • MDL number MFCD14635150
  • Cas Number : 232940-60-2
  • Molecular Weight : 172.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form solid
Country of Origin Ukraine
Availability details for
Quantity
  • Availability
  • Add to Cart